Learn More
Polysciences, Inc. Cibacron Blue, F3GA, C.I. 61211, Affinity Chromatography Grade
SDP
Catalog No. NC1730590
Encompass_Preferred
Cibacron Blue 3GA (CB 3GA) is a ortho-sulfonated-phenylamino-triazine dye that can be immobilized on a support matrix and used for affinity chromatography of proteins. Also used for probing nucleotide binding sites in proteins.
Catalog No. NC1730590 Supplier Polysciences, Inc. Supplier No. 257211
Cibacron Blue 3GA (CB 3GA) is a ortho-sulfonated-phenylamino-triazine dye that can be immobilized on a support matrix and used for affinity chromatography of proteins. Also used for probing nucleotide binding sites in proteins.