Learn More
Medchemexpress LLC Cibinetide (ARA290) | 1208243-50-8 | >99.9% | 1257.31 | 10 MG
SDP
Catalog No. 5000426068
Encompass_Preferred
Cibinetide (ARA290) is an erythropoietin (EPO) derivative that acts as a specific agonist of the erythropoietin/CD131 heteroreceptor. It is investigated for neurological disorders and has shown promise in enhancing endothelial colony-forming cell functions and attenuating EAE severity in rats. This EPO-analog peptide is notable for its potential neurotrophic and antidepressant effects without hematopoietic side-effects.
  • Acts as a specific agonist of the erythropoietin/CD131 heteroreceptor
  • Enhances proliferation, migration, and resistance to H2O2-induced apoptosis of endothelial colony-forming cells
  • May have neurotrophic and antidepressant effects
  • Prevents progressive worsening of glucose control in rats
  • Delays onset, decreases neurologic severity, and shortens duration of EAE in a dose-dependent way
Catalog No. 50-004-26068 Supplier Medchemexpress LLC Supplier No. HYP016810MG
Cibinetide (ARA290) is an erythropoietin (EPO) derivative that acts as a specific agonist of the erythropoietin/CD131 heteroreceptor. It is investigated for neurological disorders and has shown promise in enhancing endothelial colony-forming cell functions and attenuating EAE severity in rats. This EPO-analog peptide is notable for its potential neurotrophic and antidepressant effects without hematopoietic side-effects.
  • Acts as a specific agonist of the erythropoietin/CD131 heteroreceptor
  • Enhances proliferation, migration, and resistance to H2O2-induced apoptosis of endothelial colony-forming cells
  • May have neurotrophic and antidepressant effects
  • Prevents progressive worsening of glucose control in rats
  • Delays onset, decreases neurologic severity, and shortens duration of EAE in a dose-dependent way