Learn More
Aviva Systems Biology Corp CIC Antibody - N-terminal region (ARP32498_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32498_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CIon Channel. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CIC. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54625