Learn More
Cayman Chemical Ciclesonide, Purity: ≥98%, CAS: 126544-47-6, 10 mg
SDP
Catalog No. 502979806
Encompass_Preferred
A prodrug form of desisobutyryl-ciclesonide; binds to the glucocorticoid receptor (Ki = 37 nM); selective for the glucocorticoid receptor over the estrogen, progesterone, and testosterone receptors; inhibits eosinophil influx into the lung (ED50 = 0.49 mg/kg) and reduces sephadex-induced lung edema (ED50 = 0.72 mg/kg) in a rat model of ovalbumin-sensitized and -challenged airway inflammation
Catalog No. 50-297-9806 Supplier Cayman Chemical Supplier No. 1822710 MG
A prodrug form of desisobutyryl-ciclesonide; binds to the glucocorticoid receptor (Ki = 37 nM); selective for the glucocorticoid receptor over the estrogen, progesterone, and testosterone receptors; inhibits eosinophil influx into the lung (ED50 = 0.49 mg/kg) and reduces sephadex-induced lung edema (ED50 = 0.72 mg/kg) in a rat model of ovalbumin-sensitized and -challenged airway inflammation