Learn More
Medchemexpress LLC HY-B0625 50mg Medchemexpress, Ciclesonide CAS:126544-47-6 Purity:>98%
SDP
Catalog No. 502082624
Encompass_Preferred
Medchemexpress, HY-B0625 50mg Ciclesonide CAS:126544-47-6 Ciclesonide (RPR251526) is a glucocorticoid with an potent anti-inflammatory activity. Ciclesonide can be used for asthma research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2624 Supplier Medchemexpress LLC Supplier No. HYB062550MG
Medchemexpress, HY-B0625 50mg Ciclesonide CAS:126544-47-6 Ciclesonide (RPR251526) is a glucocorticoid with an potent anti-inflammatory activity. Ciclesonide can be used for asthma research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.