Learn More
Medchemexpress LLC Ciclopirox | 29342-05-0 | 98.3% | 50 MG
SDP
Catalog No. 501874104
Encompass_Preferred
Ciclopirox is a synthetic and orally active antifungal agent primarily used for superficial mycoses research. This broad-spectrum agent effectively inhibits dermatophytes, yeasts, molds, and various pathogenic Gram-positive and Gram-negative species. Beyond its antifungal properties, Ciclopirox also demonstrates anticancer and anti-inflammatory effects. It is intended for research use only.
  • Synthetic and orally active antifungal agent
  • Broad spectrum of activity against dermatophytes, yeasts, molds, and many pathogenic bacteria
  • Exhibits anticancer and anti-inflammatory effects
  • Inhibits HUVEC proliferation and angiogenesis
  • Induces cell aerobic glycolysis
  • Impairs mitochondrial functions and enhances ROS generation
  • Activates PERK-dependent ER stress
  • Reduces tumor size in mouse xenograft models
  • Inhibits tumor growth in human breast cancer xenografts in mice
Catalog No. 50-187-4104 Supplier Medchemexpress LLC Supplier No. HYB045050MG
Ciclopirox is a synthetic and orally active antifungal agent primarily used for superficial mycoses research. This broad-spectrum agent effectively inhibits dermatophytes, yeasts, molds, and various pathogenic Gram-positive and Gram-negative species. Beyond its antifungal properties, Ciclopirox also demonstrates anticancer and anti-inflammatory effects. It is intended for research use only.
  • Synthetic and orally active antifungal agent
  • Broad spectrum of activity against dermatophytes, yeasts, molds, and many pathogenic bacteria
  • Exhibits anticancer and anti-inflammatory effects
  • Inhibits HUVEC proliferation and angiogenesis
  • Induces cell aerobic glycolysis
  • Impairs mitochondrial functions and enhances ROS generation
  • Activates PERK-dependent ER stress
  • Reduces tumor size in mouse xenograft models
  • Inhibits tumor growth in human breast cancer xenografts in mice