Learn More
Apexbio Technology LLC Ciclopirox ethanolamine,50mg CAS# 41621-49-2
SDP
Catalog No. 502240443
Encompass_Preferred
Ciclopirox ethanolamine, a broad-spectrum antifungal agent, inhibits dermatophytes and yeasts pathogenic with a minimal inhibitory concentration (MIC) of 0.98-3.9 μg/mL.Ciclopirox ethanolamine, working as an iron chelator, suppresses a substantial number of clinically relevant dermatophytes, yeasts, and molds, including the frequently azole-resistant Candida species Candida glabrata, Candida krusei, and Candida guilliermondii. Moreover, Ciclopirox has been proved to inhibit a wide range of bacteria in humans, including many gram-positive and gram-negative species pathogenic bacteria. [1] Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0443 Supplier Apexbio Technology LLC Supplier No. B138450
Ciclopirox ethanolamine, a broad-spectrum antifungal agent, inhibits dermatophytes and yeasts pathogenic with a minimal inhibitory concentration (MIC) of 0.98-3.9 μg/mL.Ciclopirox ethanolamine, working as an iron chelator, suppresses a substantial number of clinically relevant dermatophytes, yeasts, and molds, including the frequently azole-resistant Candida species Candida glabrata, Candida krusei, and Candida guilliermondii. Moreover, Ciclopirox has been proved to inhibit a wide range of bacteria in humans, including many gram-positive and gram-negative species pathogenic bacteria. [1] Other sizes are also available. Please inqury us for quote.