Learn More
Medchemexpress LLC Ciclopirox (olamine) | 41621-49-2 | 99.8% | 50 MG
SDP
Catalog No. 5000408453
Encompass_Preferred
Ciclopirox olamine is a synthetic, orally active antifungal agent for superficial mycoses research. It exhibits a broad spectrum of activity against dermatophytes, yeasts, molds, and various Gram-positive and Gram-negative pathogenic species. This compound also demonstrates anticancer and anti-inflammatory effects.
  • Inhibits dermatophytes, yeasts, and molds
  • Active against Gram-positive and Gram-negative species
  • Shows anticancer activity in various cell lines and xenograft models
  • Exhibits anti-inflammatory effects
  • Inhibits HUVEC proliferation and angiogenesis
  • Induces cell aerobic glycolysis and impairs mitochondrial functions
Catalog No. 50-004-08453 Supplier Medchemexpress LLC Supplier No. HYB0450A50MG
Ciclopirox olamine is a synthetic, orally active antifungal agent for superficial mycoses research. It exhibits a broad spectrum of activity against dermatophytes, yeasts, molds, and various Gram-positive and Gram-negative pathogenic species. This compound also demonstrates anticancer and anti-inflammatory effects.
  • Inhibits dermatophytes, yeasts, and molds
  • Active against Gram-positive and Gram-negative species
  • Shows anticancer activity in various cell lines and xenograft models
  • Exhibits anti-inflammatory effects
  • Inhibits HUVEC proliferation and angiogenesis
  • Induces cell aerobic glycolysis and impairs mitochondrial functions