Learn More
TARGETMOL CHEMICALS INC Ciclopirox olamine 500MG
SDP
Catalog No. 502877587
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Ciclopirox olamine is a broad-spectrum antifungal agent with additional antibacterial and anti-inflammatory activities. Purity 98.97%
Catalog No. 50-287-7587 Supplier TARGETMOL CHEMICALS INC Supplier No. T1114500MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Ciclopirox olamine is a broad-spectrum antifungal agent with additional antibacterial and anti-inflammatory activities. Purity 98.97%