Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ciclopirox olamine

Catalog No. AC463440010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463440010 1 g
AC463440050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC463440010 Supplier Thermo Scientific Chemicals Supplier No. 463440010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41621-49-2
Molecular Formula C14H24N2O3
Molecular Weight (g/mol) 268.36
InChI Key MBRHNTMUYWQHMR-UHFFFAOYSA-N
IUPAC Name 2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one
SMILES NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1

Specifications

Melting Point 143.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: almost insoluble in dimethylsulfoxide and methanol
Formula Weight 268.35
Chemical Name or Material Ciclopirox olamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Flammable solid.

May cause fire or explosion; strong oxidizer.

Causes severe skin burns and eye damage.

May cause an allergic skin reaction.

May cause respiratory irritation.

Very toxic to aquatic life
Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

Take any precaution to avoid mixing with combustibles.

In case of major fire and large quantities: Evacuate area.

Fight fire remotely due to the risk of explosion.

Wear protective gloves/protective clothing/eye protection/face protection.

IF SWALLOWED: Rinse mouth.

Do NOT induce vomiting.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 255-464-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.