Learn More
Description
Ciclopirox Olamine USP is a white to slightly yellowish-white, crystalline powder.
Specifications
Specifications
| Chemical Name or Material | Ciclopirox Olamine USP |
| Packaging | Jar 300ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT |
| CAS | 41621-49-2 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 268.3546 |
| Quantity | 100 g |
| Percent Purity | 97.5 to 101.5 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
