Learn More
Cayman Chemical CID-1067700, Purity: ≥95%, CAS: 314042-01-8, 10 mg
SDP
Catalog No. 502971601
Encompass_Preferred
A competitive inhibitor of nucleotide binding by Ras-related GTPases; binds Rab7 (Ki = 13 nM), preventing BODIPY-linked GTP and GDP binding with EC50 values of 11.2 and 21 nM, respectively
Catalog No. 50-297-1601 Supplier Cayman Chemical Supplier No. 1519410 MG
A competitive inhibitor of nucleotide binding by Ras-related GTPases; binds Rab7 (Ki = 13 nM), preventing BODIPY-linked GTP and GDP binding with EC50 values of 11.2 and 21 nM, respectively