Learn More
Cayman Chemical CID-1375606 10mg
SDP
Catalog No. 502988885
Encompass_Preferred
A GPR27 agonist (EC50s = 0.46 µM and 7.9 µM in chimeric GPR27V2 and wild-type GPR27 luciferase b-arrestin 2 complementation assays, respectively); does not induce recruitment of b-arrestin 2 to the closely related receptors GPR85 or GPR173
Catalog No. 50-298-8885 Supplier Cayman Chemical Supplier No. 3042210 MG
A GPR27 agonist (EC50s = 0.46 µM and 7.9 µM in chimeric GPR27V2 and wild-type GPR27 luciferase b-arrestin 2 complementation assays, respectively); does not induce recruitment of b-arrestin 2 to the closely related receptors GPR85 or GPR173