Learn More
Medchemexpress LLC HY-114146 10mg Medchemexpress, CID 1375606 CAS:313493-80-0 Purity:>98%
SDP
Catalog No. 501969288
Encompass_Preferred
Medchemexpress, HY-114146 10mg CID 1375606 CAS:313493-80-0 CID 1375606 is a surrogate agonist of orphan G protein-coupled receptor GPR27. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9288 Supplier Medchemexpress LLC Supplier No. HY11414610MG
Medchemexpress, HY-114146 10mg CID 1375606 CAS:313493-80-0 CID 1375606 is a surrogate agonist of orphan G protein-coupled receptor GPR27. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.