Learn More
Cayman Chemical CID-16020046 10mg
SDP
Catalog No. 502972969
Encompass_Preferred
A selective GPR55 inverse agonist that antagonizes GPR55 constitutive activity (IC50 = 15 μM); blocks GPR55-mediated endothelial wound healing and reverses LPI-inhibited platelet aggregation
Catalog No. 50-297-2969 Supplier Cayman Chemical Supplier No. 1524710 MG
A selective GPR55 inverse agonist that antagonizes GPR55 constitutive activity (IC50 = 15 μM); blocks GPR55-mediated endothelial wound healing and reverses LPI-inhibited platelet aggregation