Learn More
Cayman Chemical CID-2745687, Purity: ≥95%, CAS: 264233-05-8, 50 mg
SDP
Catalog No. 502953764
Encompass_Preferred
A reversible, competitive antagonist of GPR35, blocking activation by the synthetic agonist pamoic acid with a Ki of 12.8 nM; less potently blocks activation of GPR35 by zaprinast (IC50 = 160 nM); shows ~57-fold selectivity for GPR35 over the related receptor GPR55 (IC50 = 9.08 µM)
Catalog No. 50-295-3764 Supplier Cayman Chemical Supplier No. 1204650 MG
A reversible, competitive antagonist of GPR35, blocking activation by the synthetic agonist pamoic acid with a Ki of 12.8 nM; less potently blocks activation of GPR35 by zaprinast (IC50 = 160 nM); shows ~57-fold selectivity for GPR35 over the related receptor GPR55 (IC50 = 9.08 µM)