Learn More
Cayman Chemical CID-2818500, Purity: ≥98%, CAS: 1215-07-2, 50 mg
SDP
Catalog No. 502964166
Encompass_Preferred
An inhibitor of PRMT1 (IC50 = 11 μM in a histone H4 methylation assay); inhibits histone H4 methylation by PRMT8, but not histone H3.1 methylation by CARM1 or Set7/9, at 10 and 100 μM
Catalog No. 50-296-4166 Supplier Cayman Chemical Supplier No. 2191950 MG
An inhibitor of PRMT1 (IC50 = 11 μM in a histone H4 methylation assay); inhibits histone H4 methylation by PRMT8, but not histone H3.1 methylation by CARM1 or Set7/9, at 10 and 100 μM