Learn More
Cayman Chemical CID-44216842 10mg
SDP
Catalog No. 502916020
Encompass_Preferred
A Cdc42 inhibitor; inhibits GTP binding to wild-type Cdc42 and Cdc42Q61L (EC50s = 0.3 and 0.5 µM, respectively)
Catalog No. 50-291-6020 Supplier Cayman Chemical Supplier No. 3777510 MG
A Cdc42 inhibitor; inhibits GTP binding to wild-type Cdc42 and Cdc42Q61L (EC50s = 0.3 and 0.5 µM, respectively)