Learn More
Cayman Chemical CID-5157334 10mg
SDP
Catalog No. 502943347
Encompass_Preferred
A surrogate agonist of GPR27; induces recruitment of β-arrestin-2 in complementation assays using wild-type GPR27 or a GPR27V2 chimeric receptor (EC50s = 7.943 and 0.417 nM, respectively) but does not induce β-arrestin-1 recruitment in the same assay or activation of the Gq, Gs, or Gi signaling pathways in aequorin-GFP assays; selective for GPR27 over GPR85V2 and GPR173V2 chimeric receptors at 15 µM
Catalog No. 50-294-3347 Supplier Cayman Chemical Supplier No. 4167710 MG
A surrogate agonist of GPR27; induces recruitment of β-arrestin-2 in complementation assays using wild-type GPR27 or a GPR27V2 chimeric receptor (EC50s = 7.943 and 0.417 nM, respectively) but does not induce β-arrestin-1 recruitment in the same assay or activation of the Gq, Gs, or Gi signaling pathways in aequorin-GFP assays; selective for GPR27 over GPR85V2 and GPR173V2 chimeric receptors at 15 µM