Learn More
TARGETMOL CHEMICALS INC CID-663143 5MG
SDP
Catalog No. 502580840
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CID-663143 inhibits cancer cell growth and shows microtubule-binding activity. purity: 99%
Catalog No. 50-258-0840 Supplier TARGETMOL CHEMICALS INC Supplier No. T238895MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CID-663143 inhibits cancer cell growth and shows microtubule-binding activity. purity: 99%