Learn More
Selleck Chemical LLC CID16020046 5mg 834903-43-4 C390-0219
SDP
Supplier:  Selleck Chemical LLC S86945MG
Encompass_Preferred
CID16020046 (C390-0219) is a selective GPR55 antagonist, inhibiting GPR55 constitutive activity with IC50 of 0.15 ?M in yeast. It demonstrates weak activity against a broad spectrum of other GPCRs, ion channels, kinases, and nuclear receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8366