Learn More
Medchemexpress LLC Cid44216842 (Cdc42-IN-1) | 1222513-26-9 | 99.8% | C22H20BrN3O3S | 10MG
SDP
Catalog No. 5000261692
Encompass_Preferred
CID44216842 (Cdc42-IN-1) is a potent, selective small-molecule inhibitor of the Rho GTPase Cdc42 that blocks GTP and GDP binding to both wild-type and Q61L mutant forms. Supplied as a solid white to off-white powder with high purity, it is intended for use as a molecular probe in biochemical and cellular research.
  • Selective inhibition of Cdc42 over other GTPases.
  • Low-micromolar potency in GTP and GDP binding assays.
  • High chemical purity suitable for research applications.
  • Stable solid form with defined storage conditions.
  • Useful as a molecular probe for studying Cdc42-mediated signaling.
Catalog No. 50-002-61692 Supplier Medchemexpress LLC Supplier No. HY13637910MG
CID44216842 (Cdc42-IN-1) is a potent, selective small-molecule inhibitor of the Rho GTPase Cdc42 that blocks GTP and GDP binding to both wild-type and Q61L mutant forms. Supplied as a solid white to off-white powder with high purity, it is intended for use as a molecular probe in biochemical and cellular research.
  • Selective inhibition of Cdc42 over other GTPases.
  • Low-micromolar potency in GTP and GDP binding assays.
  • High chemical purity suitable for research applications.
  • Stable solid form with defined storage conditions.
  • Useful as a molecular probe for studying Cdc42-mediated signaling.