Learn More
Medchemexpress LLC HY-100502 5mg Medchemexpress, CID5721353 CAS:301356-95-6 Purity:>98%
SDP
Catalog No. 501931858
Encompass_Preferred
Medchemexpress, HY-100502 5mg CID5721353 CAS:301356-95-6 CID5721353 is an inhibitor of BCL6 with an IC 50 value of 212 μM, which corresponds to a K i of 147 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1858 Supplier Medchemexpress LLC Supplier No. HY1005025MG
Medchemexpress, HY-100502 5mg CID5721353 CAS:301356-95-6 CID5721353 is an inhibitor of BCL6 with an IC 50 value of 212 μM, which corresponds to a K i of 147 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.