Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-hydroxy-2,3,4,5-tetrahydrobenzofuro[2,3-c]azepin-1-one | 521937-07-5 | 99.1% | 5 MG
SDP

Supplier:  Medchemexpress LLC HY122395MG

Encompass_Preferred

CID755673 is a small-molecule inhibitor of protein kinase D (PKD) with reported IC50 values for PKD1, PKD2, and PKD3. It is supplied as a high-purity solid for research use and is suitable for biochemical, cellular, and in vivo studies.

  • Potent PKD inhibition: PKD1 IC50 182 nM, PKD2 IC50 280 nM, PKD3 IC50 227 nM.
  • High purity solid suitable for biochemical and cellular assays.
  • Soluble in DMSO at 100 mg/mL for in vitro applications.
  • Recommended storage: powder at -20°C for long-term stability.
  • In vivo dosing examples reported: 1 or 10 mg/kg intraperitoneal in mice.

Catalog No. 50-208-0984


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.