Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decon Labs, Inc. CiDecon™ II

Catalog No. 18800111 Shop All Decon Labs Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
18-800-111 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-800-111 Supplier Decon Labs, Inc. Supplier No. 8512

missing translation for 'validMainframeMsg'

Single serve sterile, gamma-irradiated, concentrated phenolic disinfectant

  • Phosphate-free detergent enhanced disinfectant
  • For use in cleanroom areas and surfaces where broad-based disinfection is required
  • Designed to clean, disinfect and deodorize any washable, inanimate, non-porous surface in one easy step
  • Can be alternated with LopHene II ST if rotational disinfection program is desired or where organism resistance is suspected
  • Each lot tested for sterility USP 14-day sterility test prior to release
  • Dilution of 1 oz./gal.
  • Used with two gal. of water
  • Sterility assurance level: 10-6
  • Double-bagged

Specifications

Product Type EPA Registered Disinfectant
pH 12 to 13
Sterility Sterile
Container Type Double-bagged Bottles
For Use With (Application) For use in cleanroom areas and surfaces where broad-based disinfection is required
Includes Lot specific document
Quantity 24 Pk.
Volume (English) 2 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.