Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decon™ CiDecon Plus Wipes

Catalog No. 0435587 Shop All Decon Labs Products
Change view
Click to view available options
Dimensions (L x W):
5 x 8 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
04-355-87 5 x 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 04-355-87 Supplier Decon™ Supplier No. 8511

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Premoistened phenolic disinfectant wipes for use on hard surfaces

  • Achieve hospital disinfection in three minutes and maintain continual residual bacteriostatic activity for up to six months
  • Economical, durable wipes display superior efficacy against a broad range of pathogenic organisms
  • Tuberculocidal, virucidal, bactericidal, and fungicidal

Compliance

Meet OSHA Bloodborne Pathogen Standard. EPA registered.

Specifications

Product Type Disinfectant Wipe
Length (English) 5 in.
Length (Metric) 12.7 cm
Width (English) 4.5 in.
Width (Metric) 11.43 cm
Certifications/Compliance FDA, OSHA, EPA
Container Type Pop-up Canister
Dimensions (L x W) 5 x 8 in.
For Use With (Application) Efficacy against a broad range of pathogenic organisms on hard surfaces
Packaging Quantity 180 Wipes/Container
Saturation Solution 1.56% Buffered Phenol, 0.06% Sodium Phenate
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.