Learn More
Medchemexpress LLC Cidofovir diphosphate tri(triethylamine) | 00-00-0 | 99.9% | 742.72 g/mol | C26H61N6O12P3 | 5 MG
SDP
Catalog No. 5000262575
Encompass_Preferred
Cidofovir diphosphate tri(triethylamine) is the triethylamine salt of cidofovir diphosphate, an active intracellular metabolite and selective inhibitor of viral DNA polymerases. It is provided as a high-purity biochemical reagent intended for in vitro and biochemical research applications.
Catalog No. 50-002-62575 Supplier Medchemexpress LLC Supplier No. HY131606B5MG
Cidofovir diphosphate tri(triethylamine) is the triethylamine salt of cidofovir diphosphate, an active intracellular metabolite and selective inhibitor of viral DNA polymerases. It is provided as a high-purity biochemical reagent intended for in vitro and biochemical research applications.