Learn More
Selleck Chemical LLC Cidofovir 100mg 113852-37-2 HPMPC
SDP
Supplier:  Selleck Chemical LLC S1516100MG
Encompass_Preferred
Cidofovir (HPMPC) suppresses virus replication by selective inhibition of viral DNA synthesis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-791-2