Learn More
Grainger Cigarette Receptacle 4 gal Silver
Catalog No. 502749551
Encompass_Preferred
The Justrite Replacement Pail is a 5 gallon steel pail with a silver powder coat finish. It has a leakproof lid with a gasket seal and 3/4" threaded opening. The pail meets OSHA and NFPA requirements for the storage of flammable liquids. It can be used with Justrite safety cans for the safe handling, storage and dispensing of flammables. This UN rated pail has rolled edges for added strength.
Catalog No. 50-274-9551 Supplier Grainger Supplier No. 14N878
The Justrite Replacement Pail is a 5 gallon steel pail with a silver powder coat finish. It has a leakproof lid with a gasket seal and 3/4" threaded opening. The pail meets OSHA and NFPA requirements for the storage of flammable liquids. It can be used with Justrite safety cans for the safe handling, storage and dispensing of flammables. This UN rated pail has rolled edges for added strength.