Learn More
Medchemexpress LLC Cigb-300 (acetate) | 98.0% | 3060.60 | 1 MG
SDP
Catalog No. 5000400205
Encompass_Preferred
CIGB-300 acetate (P15-Tat acetate) is an anti-casein kinase 2 (CK2) peptide that exhibits anticancer properties by interfering with the phosphorylation of protein kinase CK2. CIGB-300 acetate induces apoptosis in multiple tumor cell lines and can be used in cancer research.
  • Exhibits anticancer properties
  • Interferes with phosphorylation of protein kinase CK2
  • Induces apoptosis in multiple tumor cell lines
  • Useful in cancer research
Catalog No. 50-004-00205 Supplier Medchemexpress LLC Supplier No. HYP4056A1MG
CIGB-300 acetate (P15-Tat acetate) is an anti-casein kinase 2 (CK2) peptide that exhibits anticancer properties by interfering with the phosphorylation of protein kinase CK2. CIGB-300 acetate induces apoptosis in multiple tumor cell lines and can be used in cancer research.
  • Exhibits anticancer properties
  • Interferes with phosphorylation of protein kinase CK2
  • Induces apoptosis in multiple tumor cell lines
  • Useful in cancer research