Learn More
Cayman Chemical Ciglitazone, Purity: ≥98%, CAS: 74772-77-3, 100 mg
SDP
Catalog No. 502932458
Encompass_Preferred
An antidiabetic drug and selective PPARγ agonist (EC50 = 3.0 µM); active in vivo as an anti-hyperglycemic agent the ob/ob mouse model
Catalog No. 50-293-2458 Supplier Cayman Chemical Supplier No. 71730100 MG
An antidiabetic drug and selective PPARγ agonist (EC50 = 3.0 µM); active in vivo as an anti-hyperglycemic agent the ob/ob mouse model