Learn More
AdipoGen Ciglitazone (25 mg)
SDP
Supplier:  AdipoGen AGCR10033M025
Encompass_Preferred
Chemical. CAS: 74772-77-3. Formula: C18H23NO3S. MW: 333.5. Selective PPARgamma agonist (EC50=3 µM). Hypoglycemic agent. Angiogenesis inhibitor. Anti-inflammatory. Apoptosis inducer.
Catalog No. 50-205-3788