Learn More
Abcam Ciglitazone, PPARgamma agonist, 10MG
Catalog No. 502706858
Encompass_Preferred
MW 333.4 Da, Purity >98%. Selective PPARγ agonist (EC₅₀ = 3 μM). Shows antidiabetic and antineoplastic activity. Induces cell cycle arrest and apoptosis. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6858 Supplier Abcam Supplier No. AB14113910MG
MW 333.4 Da, Purity >98%. Selective PPARγ agonist (EC₅₀ = 3 μM). Shows antidiabetic and antineoplastic activity. Induces cell cycle arrest and apoptosis. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement