Learn More
Medchemexpress LLC HY-112063 5mg Medchemexpress, CIL56 CAS:300802-28-2 Purity:>98%
SDP
Catalog No. 501969008
Encompass_Preferred
Medchemexpress, HY-112063 5mg CIL56 CAS:300802-28-2 CIL56 is a potent and selective ferroptosis inducer. Ferroptosis is an iron-dependent form of regulated cell death (RCD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9008 Supplier Medchemexpress LLC Supplier No. HY1120635MG
Medchemexpress, HY-112063 5mg CIL56 CAS:300802-28-2 CIL56 is a potent and selective ferroptosis inducer. Ferroptosis is an iron-dependent form of regulated cell death (RCD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.