Learn More
Medchemexpress LLC CIL62 | 117593-36-9 | 97.0% | 382.45 | 100 MG
SDP
Catalog No. 5000351236
Encompass_Preferred
CIL62 is a caspase-3/7-independent cell death inducer. Its mechanism of action is Necrostatin-1 dependent cell death.
  • COLO 205: IC50 28 μM (Compound: 3f) for antiproliferative activity after 24 hrs by MTT assay.
  • IMR-32: IC50 41 μM (Compound: 3f) for antiproliferative activity after 24 hrs by MTT assay.
  • K562: IC50 36 μM (Compound: 3f) for antiproliferative activity after 24 hrs by MTT assay.
  • Necrostatin-1 suppresses CIL62 at 19 μM (5 μg/mL).
Catalog No. 50-003-51236 Supplier Medchemexpress LLC Supplier No. HY136509100MG
CIL62 is a caspase-3/7-independent cell death inducer. Its mechanism of action is Necrostatin-1 dependent cell death.
  • COLO 205: IC50 28 μM (Compound: 3f) for antiproliferative activity after 24 hrs by MTT assay.
  • IMR-32: IC50 41 μM (Compound: 3f) for antiproliferative activity after 24 hrs by MTT assay.
  • K562: IC50 36 μM (Compound: 3f) for antiproliferative activity after 24 hrs by MTT assay.
  • Necrostatin-1 suppresses CIL62 at 19 μM (5 μg/mL).