Learn More
Medchemexpress LLC CIL62 | 117593-36-9 | 97.0% | 382.45 | 50 MG
SDP
Catalog No. 5000350244
Encompass_Preferred
CIL62 is a caspase-3/7-independent cell death inducer that operates via Necrostatin-1 dependent mechanisms. It exhibits antiproliferative activity against human COLO205, IMR32, and K562 cells.
  • Caspase-3/7-independent cell death inducer
  • Necrostatin-1 dependent mechanism of action
  • Antiproliferative activity against COLO205, IMR32, and K562 cells
Catalog No. 50-003-50244 Supplier Medchemexpress LLC Supplier No. HY13650950MG
CIL62 is a caspase-3/7-independent cell death inducer that operates via Necrostatin-1 dependent mechanisms. It exhibits antiproliferative activity against human COLO205, IMR32, and K562 cells.
  • Caspase-3/7-independent cell death inducer
  • Necrostatin-1 dependent mechanism of action
  • Antiproliferative activity against COLO205, IMR32, and K562 cells