Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cil62 | 117593-36-9 | 97.0% | C23H26O5 | 10MG
SDP

Catalog No. 5000259754
Encompass_Preferred

CIL62 is a small-molecule research compound that induces caspase-3/7-independent cell death through a necrostatin-1-dependent pathway. It is supplied as a purified powder for laboratory research use only; consult the datasheet and SDS for handling, stability, and safety information.

  • Induces caspase-3/7-independent cell death via a necrostatin-1-dependent mechanism.
  • Molecular formula C23H26O5; molecular weight 382.45 g/mol.
  • Purity 97.0% by HPLC.
  • Available as a powder in multiple package sizes for laboratory research.
  • Recommended storage: powder at -20°C for long-term, 4°C for short-term.

Catalog No. 50-002-59754 Supplier Medchemexpress LLC Supplier No. HY13650910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CIL62 is a small-molecule research compound that induces caspase-3/7-independent cell death through a necrostatin-1-dependent pathway. It is supplied as a purified powder for laboratory research use only; consult the datasheet and SDS for handling, stability, and safety information.

  • Induces caspase-3/7-independent cell death via a necrostatin-1-dependent mechanism.
  • Molecular formula C23H26O5; molecular weight 382.45 g/mol.
  • Purity 97.0% by HPLC.
  • Available as a powder in multiple package sizes for laboratory research.
  • Recommended storage: powder at -20°C for long-term, 4°C for short-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.