Learn More
TARGETMOL CHEMICALS INC CILASTATIN 100MG
SDP
Catalog No. 502577089
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cilastatin (MK0791) is a renal dehydropeptidase-I and leukotriene D4 dipeptidase inhibitor. Since the antibiotic IMIPENEM is hydrolyzed by dehydropeptidase-I which resides in the brush border of the renal tubule cilastatin is administered with imipenem to increase its effectiveness. The drug also inhibits the metabolism of leukotriene D4 to leukotriene E4. purity: 99%
Catalog No. 50-257-7089 Supplier TARGETMOL CHEMICALS INC Supplier No. T2583100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cilastatin (MK0791) is a renal dehydropeptidase-I and leukotriene D4 dipeptidase inhibitor. Since the antibiotic IMIPENEM is hydrolyzed by dehydropeptidase-I which resides in the brush border of the renal tubule cilastatin is administered with imipenem to increase its effectiveness. The drug also inhibits the metabolism of leukotriene D4 to leukotriene E4. purity: 99%