Learn More
Cayman Chemical Cilastatin-15N-d3, Purity: ≥99% deuterated forms(d1-d3); ≤1% d0, CAS: 2738376-83-3, 1 mg
SDP
Catalog No. 502940426
Encompass_Preferred
An internal standard for the quantification of cilastatin by GC- or LC-MS
Catalog No. 50-294-0426 Supplier Cayman Chemical Supplier No. 290981 MG
An internal standard for the quantification of cilastatin by GC- or LC-MS