Learn More
Cayman Chemical CIlastatIn-15N-d3 500ug
SDP
Catalog No. 502937510
Encompass_Preferred
An internal standard for the quantification of cilastatin by GC- or LC-MS
Catalog No. 50-293-7510 Supplier Cayman Chemical Supplier No. 29098500 UG
An internal standard for the quantification of cilastatin by GC- or LC-MS