Learn More
Enzo Life Sciences Cilastatin Sodium (50mg). CAS: 81129-83-1
SDP
Catalog No. 502011491
Encompass_Preferred
Cilastatin is a brush border dipeptidase inhibitor. It inhibits the conversion of leukotriene D4 to leukotriene E4. Nephroprotective in cyclosporine A-induced nephrotoxicity. Alternative name: [R-[R*,S*-(Z)]]-7-[(2-Amino-2-carboxyethyl)thio]-2- [[(2,2-dimethylcyclopropyl)carbonyl]amino]-2- heptenoic acid monosodium salt. Purity: ≥98%. Solubility: Soluble in water (25mg/ml) or DMSO (10mg/ml at 37°C). Long Term Storage: -20°C.
Catalog No. 50-201-1491 Supplier Enzo Life Sciences Supplier No. BMLPI1530050
Cilastatin is a brush border dipeptidase inhibitor. It inhibits the conversion of leukotriene D4 to leukotriene E4. Nephroprotective in cyclosporine A-induced nephrotoxicity. Alternative name: [R-[R*,S*-(Z)]]-7-[(2-Amino-2-carboxyethyl)thio]-2- [[(2,2-dimethylcyclopropyl)carbonyl]amino]-2- heptenoic acid monosodium salt. Purity: ≥98%. Solubility: Soluble in water (25mg/ml) or DMSO (10mg/ml at 37°C). Long Term Storage: -20°C.