Learn More
Medchemexpress LLC Cilazapril (monohydrate) | 92077-78-6 | 99.9% | C22H33N3O6 | 25 MG
SDP
Catalog No. 5000357830
Encompass_Preferred
Cilazapril monohydrate is an orally active prodrug of the angiotensin-converting enzyme (ACE) inhibitor Cilazaprilat. It reduces plasma ACE activity and is used in the research of hypertension (including essential and renal hypertension) and congestive heart failure.
  • Reduces plasma ACE activity
  • Used in research of hypertension and congestive heart failure
  • Evokes similar maximum degree of plasma ACE inhibition as Enalapril
  • Dose-dependently reduces plasma ACE activity and pressor response to angiotensin in cats
  • Gradually reduces blood pressure in dogs with volume-depleted renal hypertension
  • Decreases blood pressure, reduces heart weight to body weight ratio, and improves postischemic myocardial function in hyperthyroid rats
Catalog No. 50-003-57830 Supplier Medchemexpress LLC Supplier No. 25MGHYA0043A
Cilazapril monohydrate is an orally active prodrug of the angiotensin-converting enzyme (ACE) inhibitor Cilazaprilat. It reduces plasma ACE activity and is used in the research of hypertension (including essential and renal hypertension) and congestive heart failure.
  • Reduces plasma ACE activity
  • Used in research of hypertension and congestive heart failure
  • Evokes similar maximum degree of plasma ACE inhibition as Enalapril
  • Dose-dependently reduces plasma ACE activity and pressor response to angiotensin in cats
  • Gradually reduces blood pressure in dogs with volume-depleted renal hypertension
  • Decreases blood pressure, reduces heart weight to body weight ratio, and improves postischemic myocardial function in hyperthyroid rats