Learn More
Medchemexpress LLC Cilazapril monohydrate | 92077-78-6 | MFCD00865788 | 99.9% | C22H33N3O6 | 100 MG
SDP
Catalog No. 5000433471
Encompass_Preferred
Cilazapril monohydrate is an orally active prodrug of the angiotensin-converting enzyme (ACE) inhibitor Cilazaprilat. It functions by reducing plasma ACE activity. This compound can be utilized in the research of hypertension, including essential and renal hypertension, and congestive heart failure.
  • Orally active prodrug of an ACE inhibitor
  • Reduces plasma ACE activity
  • Used in research of hypertension (essential and renal)
  • Used in research of congestive heart failure
Catalog No. 50-004-33471 Supplier Medchemexpress LLC Supplier No. HYA0043A100MG
Cilazapril monohydrate is an orally active prodrug of the angiotensin-converting enzyme (ACE) inhibitor Cilazaprilat. It functions by reducing plasma ACE activity. This compound can be utilized in the research of hypertension, including essential and renal hypertension, and congestive heart failure.
  • Orally active prodrug of an ACE inhibitor
  • Reduces plasma ACE activity
  • Used in research of hypertension (essential and renal)
  • Used in research of congestive heart failure