Learn More
Medchemexpress LLC Cilazapril monohydrate | 92077-78-6 | MFCD00865788 | 99.9% | C22H33N3O6 | 5 MG
SDP
Catalog No. 5000432270
Encompass_Preferred
Cilazapril monohydrate is an orally active prodrug of the angiotensin-converting enzyme (ACE) inhibitor Cilazaprilat. It reduces plasma ACE activity and can be used in the research of hypertension (including essential and renal hypertension) and congestive heart failure.
  • Orally active prodrug of Cilazaprilat
  • Reduces plasma ACE activity
  • Applicable in research for hypertension (essential and renal) and congestive heart failure
  • Evokes a maximum degree of plasma ACE inhibition (-76%) in rats
  • Dose-dependently reduces plasma ACE activity and pressor response to angiotensin in cats
  • Gradually reduces blood pressure in dogs with volume-depleted renal hypertension
  • Decreases blood pressure, reduces heart weight to body weight ratio, and improves postischemic myocardial function in hyperthyroid rats
Catalog No. 50-004-32270 Supplier Medchemexpress LLC Supplier No. HYA0043A5MG
Cilazapril monohydrate is an orally active prodrug of the angiotensin-converting enzyme (ACE) inhibitor Cilazaprilat. It reduces plasma ACE activity and can be used in the research of hypertension (including essential and renal hypertension) and congestive heart failure.
  • Orally active prodrug of Cilazaprilat
  • Reduces plasma ACE activity
  • Applicable in research for hypertension (essential and renal) and congestive heart failure
  • Evokes a maximum degree of plasma ACE inhibition (-76%) in rats
  • Dose-dependently reduces plasma ACE activity and pressor response to angiotensin in cats
  • Gradually reduces blood pressure in dogs with volume-depleted renal hypertension
  • Decreases blood pressure, reduces heart weight to body weight ratio, and improves postischemic myocardial function in hyperthyroid rats