Learn More
Cayman Chemical CIlazaprIlat 10mg
SDP
Catalog No. 502916287
Encompass_Preferred
An ACE inhibitor (IC50 = 0.7 nM for the rat enzyme) and active metabolite of cilazapril; decreases creatine kinase release in primary neonatal rat cardiac myocytes in an in vitro model of ischemia-reperfusion injury induced by hypoxia and reoxygenation at 10 µM; increases coronary blood flow and fractional shortening in a dog model of coronary hypoperfusion-induced myocardial ischemia at 3 UG/kg per minute intra-arterially
Catalog No. 50-291-6287 Supplier Cayman Chemical Supplier No. 3811810 MG
An ACE inhibitor (IC50 = 0.7 nM for the rat enzyme) and active metabolite of cilazapril; decreases creatine kinase release in primary neonatal rat cardiac myocytes in an in vitro model of ischemia-reperfusion injury induced by hypoxia and reoxygenation at 10 µM; increases coronary blood flow and fractional shortening in a dog model of coronary hypoperfusion-induced myocardial ischemia at 3 UG/kg per minute intra-arterially