Learn More
Medchemexpress LLC HY-16141 10mg , Cilengitide CAS:188968-51-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1614110MG
Encompass_Preferred
HY-16141 10mg Cilengitide CAS: 188968-51-6 Cilengitide (EMD 121974) is a potent and selective inhibitor of the integrins ανβ3 and ανβ5. Cilengitide inhibits binding of isolated ανβ3 and ανβ5 to Vitronectin with an IC50 value of 4 and 79 nM, respectively . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9545