Learn More
Carbosynth LLC. Cilengitide, 1mg, CAS# 188968-51-6, MF= C27H40N8O7
SDP
Supplier:  Carbosynth LLC. RGD3036PI1MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Cilingitide; Cyclo(Arg-Gly-Asp-D-Phe-N-Me-Val), Application: Selective Inhibitor of v3 and v5 with Possible Role in Tumor Therapy, salt: Trifluoroacetate Salt, one letter code: c(RGDf-NMeV), MW: 588.68
Catalog No. 50-195-2429