Learn More
Carbosynth LLC. Cilengitide, 25mg, CAS# 188968-51-6, MF= C27H40N8O7
SDP
Supplier:  Carbosynth LLC. RGD3036PI25MG
Encompass_Preferred
Cilingitide; Cyclo(Arg-Gly-Asp-D-Phe-N-Me-Val), Application: Selective Inhibitor of αvß3 and αvß5 with Possible Role in Tumor Therapy, salt: Trifluoroacetate Salt, one letter code: c(RGDf-NMeV), MW: 588.68
Catalog No. 50-195-2430