Learn More
Apexbio Technology LLC Cilengitide, 50mg. Cas: 188968-51-6 MFCD: MFCD04307743. Integrin inhibitor for αvβ3 and αvβ5
SDP
Supplier:  Apexbio Technology LLC A866050
Encompass_Preferred
Cilengitide is a cyclic RGD pentapeptide [Arg-Gly-Asp-DPhe-(NMeVal)], and a potent v3 and v5 integrin inhibitor to block integrin-mediated adhesion and migration. It can directly bind v3 integrin. Its IC50 is 250 nM as an v3 inhibitor [1] [2] [3] [4]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5177