Learn More
Medchemexpress LLC Cilengitide TFA | 199807-35-7 | 99.9% | 702.68 | 25 MG
SDP
Catalog No. 5000394413
Encompass_Preferred
Cilengitide is a potent and selective integrin inhibitor for αvβ3 and αvβ5 receptor, with IC50 values of 4 nM and 79 nM, respectively. It inhibits integrin-mediated binding to vitronectin.
  • Potent and selective integrin inhibitor
  • Targets αvβ3 receptor with IC50 of 4 nM
  • Targets αvβ5 receptor with IC50 of 79 nM
Catalog No. 50-003-94413 Supplier Medchemexpress LLC Supplier No. 25MGHY16143
Cilengitide is a potent and selective integrin inhibitor for αvβ3 and αvβ5 receptor, with IC50 values of 4 nM and 79 nM, respectively. It inhibits integrin-mediated binding to vitronectin.
  • Potent and selective integrin inhibitor
  • Targets αvβ3 receptor with IC50 of 4 nM
  • Targets αvβ5 receptor with IC50 of 79 nM